2-Mercaptopyridin-N-oxid ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A14152.06 A14152.14 A14152.22
A14152.06EA 66.5 EUR
A14152.06 A14152.14 A14152.22
2-Mercaptopyridin-N-oxid ≥99%
2-Mercaptopyridin-N-oxid

Purity: 99% CAS No.: 1121-31-9 Molecular Formula: C5H5NOS Formula Weight: 127.17 Melting Point: 69-72° Beilstein Reference: 906983 Merck Reference: 14,7994 MDL No.: MFCD00006196

Formel: C₅H₅NOS
Schmelzpunkt: 69…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006196
CAS-Nummer: 1121-31-9
EINECS: 214-329-4
Merck Index: 14,7994

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR