2-Chlor-6-methylisonicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-6-methylpyridine-4-carboxylic acid

017200-2.5G 017200-1G 017200-5G
FLUO017200-2.5GEA 0 EUR
FLUO017200-2.5G FLUO017200-1G FLUO017200-5G
2-Chlor-6-methylisonicotinsäure
2-Chlor-6-methylisonicotinsäure
Formel: C₇H₆ClNO₂
Molecular Weight: 171.58 g/mol
MDL: MFCD00052830
CAS-Nummer: 25462-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR