2-Chlor-6-methylisonicotinsäure

Lieferant: Thermo Scientific

Synonyme: 2-Chloro-6-methylpyridine-4-carboxylic acid

RF03759DA RF03759EA
MAYBRF03759DAEA 0 EUR
MAYBRF03759DA MAYBRF03759EA
2-Chlor-6-methylisonicotinsäure
2-Chlor-6-methylisonicotinsäure
Formel: C₇H₆ClNO₂
Molecular Weight: 171.58 g/mol
MDL: MFCD00052830
CAS-Nummer: 25462-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR