2-Chlor-6-methylisonicotinsäure, Sigma-Aldrich®
Synonyme:
2-Chloro-6-methylpyridine-4-carboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
649457-1G
649457-5G
SIAL649457-1GEA
41.3
EUR
SIAL649457-1G
SIAL649457-5G
2-Chlor-6-methylisonicotinsäure, Sigma-Aldrich®
2-Chlor-6-methylisonicotinsäure
Formel:
C₇H₆ClNO₂ Molecular Weight: 171.58 g/mol |
MDL:
MFCD00052830 CAS-Nummer: 25462-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...