2-Chlor-6-(trifluormethyl)nicotinsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Chloro-6-(trifluoromethyl)pyridine-3-carboxylic acid
009102-1G
009102-5G
FLUO009102-1GEA
75.9
EUR
FLUO009102-1G
FLUO009102-5G
2-Chlor-6-(trifluormethyl)nicotinsäure
2-Chlor-6-(trifluormethyl)nicotinsäure
Formel:
C₇H₃ClF₃NO₂ Molecular Weight: 225.56 g/mol |
MDL:
MFCD01862658 CAS-Nummer: 280566-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...