6-Methylnicotinamid ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Methylpyridine-3-carboxamide
A16039.03
A16039.06
A16039.03EA
63.7
EUR
A16039.03
A16039.06
6-Methylnicotinamid ≥98%
6-Methylnicotinamid
Purity: 98%
CAS No.: 6960-22-1
Molecular Formula: C7H8N2O
Formula Weight: 136.15
Melting Point: 195-199°
Beilstein Reference: 112050
Formel:
C₇H₈N₂O Molecular Weight: 136,15 g/mol Schmelzpunkt: 195…199 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006342 CAS-Nummer: 6960-22-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...