3-Brom-2-chlor-5-thiophensulfonylchlorid

Lieferant: Thermo Scientific

Synonyme: 4-Bromo-5-chlorothiophene-2-sulphonyl chloride

SEW02652.0001 SEW02652CB SEW02652DA
MAYBSEW02652.0001EA 0 EUR
MAYBSEW02652.0001 MAYBSEW02652CB MAYBSEW02652DA
3-Brom-2-chlor-5-thiophensulfonylchlorid
3-Brom-2-chlor-5-thiophensulfonylchlorid
Formel: C₄HBrCl₂O₂S₂
Molecular Weight: 295,9895 g/mol
MDL: MFCD00052094
CAS-Nummer: 166964-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR