4-Brom-2-thiophencarbonitril
Lieferant: COMBI-BLOCKS
Synonyme:
4-Bromothiophene-2-carbonitrile, 4-Bromo-2-cyanothiophene
HI-1311-5G
HI-1311-25G
HI-1311-1G
COBBHI-1311-5GEA
88.6
EUR
COBBHI-1311-5G
COBBHI-1311-25G
COBBHI-1311-1G
4-Brom-2-thiophencarbonitril
4-Brom-2-thiophencarbonitril
Formel:
C₅H₂BrNS Molecular Weight: 188,0500 g/mol |
MDL:
MFCD00114951 CAS-Nummer: 18791-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...