2,5-Dichlor-4-nitro-3-thiophensulfonylchlorid

Lieferant: Thermo Scientific

CD11390.250 CD11390.1
MAYBCD11390.250EA 0 EUR
MAYBCD11390.250 MAYBCD11390.1
2,5-Dichlor-4-nitro-3-thiophensulfonylchlorid
2,5-Dichlor-4-nitro-3-thiophensulfonylchlorid
Formel: C₄Cl₃NO₄S₂
MDL: MFCD01570024
CAS-Nummer: 59768-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR