Thiophen-2-carboxamid ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Thiophenecarboxamide

A15219.06 A15219.14
A15219.06EA 50.6 EUR
A15219.06 A15219.14
Thiophen-2-carboxamid ≥99%
Thiophen-2-carboxamid

Purity: 99% CAS No.: 5813-89-8 Molecular Formula: C5H5NOS Formula Weight: 127.17 Melting Point: 178-183° Beilstein Reference: 110153

Formel: C₅H₅NOS
Molecular Weight: 127,17 g/mol
Schmelzpunkt: 178…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010425
CAS-Nummer: 5813-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR