Thiophen-2-carboxamid
Lieferant: Thermo Scientific
Synonyme:
2-Thiophenecarboxamide
BTB07541.1
BTB07541.10
BTB07541.50
MAYBBTB07541.1EA
0
EUR
MAYBBTB07541.1
MAYBBTB07541.10
MAYBBTB07541.50
Thiophen-2-carboxamid
Thiophen-2-carboxamid
Formel:
C₅H₅NOS Molecular Weight: 127,17 g/mol |
MDL:
MFCD00010425 CAS-Nummer: 5813-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...