2,5-Thiophendicarbonsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Thiophene-2,5-dicarboxylic acid

L10293.03 L10293.06 L10293.14
L10293.03EA 26 EUR
L10293.03 L10293.06 L10293.14
2,5-Thiophendicarbonsäure ≥97%
2,5-Thiophenedicarboxylic acid

Purity: 97% CAS No.: 4282-31-9 Molecular Formula: C6H4O4S Formula Weight: 172.16 Melting Point: 355-360° Beilstein Reference: 136781

Formel: C₆H₄O₄S
Molecular Weight: 172,16 g/mol
Schmelzpunkt: 355…360 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016896
CAS-Nummer: 4282-31-9
EINECS: 224-284-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR