2,5-Thiophendicarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Thiophene-2,5-dicarboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

405191-25G 405191-5G
SIAL405191-25GEA 31.8 EUR
SIAL405191-25G SIAL405191-5G
2,5-Thiophendicarbonsäure, Sigma-Aldrich®
2,5-Thiophenedicarboxylic acid
Formel: C₆H₄O₄S
Molecular Weight: 172,16 g/mol
MDL: MFCD00016896
CAS-Nummer: 4282-31-9
EINECS: 224-284-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR