2-(Methylthio)thiophen

Lieferant: Thermo Scientific

Synonyme: 2-(Mehtylthio)thiophene

KM01413.1 KM01413.10
MAYBKM01413.1EA 0 EUR
MAYBKM01413.1 MAYBKM01413.10
2-(Methylthio)thiophen
2-(Methylthio)thiophene
Formel: C₅H₆S₂
Molecular Weight: 130.23 g/mol
MDL: MFCD00052382
CAS-Nummer: 5780-36-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR