2-(Phenylthio)thiophen ≥97%

Lieferant: Thermo Scientific

A13708.03 A13708.06
A13708.03EA 46.6 EUR
A13708.03 A13708.06
2-(Phenylthio)thiophen ≥97%
2-(Phenylthio)thiophene

Purity: 97+% CAS No.: 16718-12-0 Molecular Formula: C10H8S2 Formula Weight: 192.30 Boiling Point: 109-110°/0.4mm MDL No.: MFCD00052141

Formel: C₁₀H₈S₂
Molecular Weight: 192,30 g/mol
Siedepunkt: 109…110 °C (0,4 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00052141
CAS-Nummer: 16718-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR