2-(Phenylthio)thiophen
Lieferant: Thermo Scientific
KM04712.1
KM04712.10
MAYBKM04712.1EA
0
EUR
MAYBKM04712.1
MAYBKM04712.10
2-(Phenylthio)thiophen
2-(Phenylthio)thiophene
Formel:
C₁₀H₈S₂ Molecular Weight: 192,30 g/mol |
MDL:
MFCD00052141 CAS-Nummer: 16718-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...