Thiophen-2-ethylamin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-(Thiophen-2-yl)ethanamine, 2-(2-Aminoethyl)thiophene

A10920.14 A10920.03 A10920.06
A10920.14EA 434 EUR
A10920.14 A10920.03 A10920.06
Thiophen-2-ethylamin 98%
Thiophen-2-ethylamin

Purity: 98% CAS No.: 30433-91-1 Molecular Formula: C6H9NS Formula Weight: 127.21 Storage Temperature: 2°C to 8°C Flash Point: 87°(188°F) Refractive Index: 1.5510 Boiling Point: 201° Beilstein Reference: 106962 UN No.: 2735 MDL No.: MFCD00051495

Formel: C₆H₉NS
Molecular Weight: 127.21 g/mol
Siedepunkt: 201 °C
Dichte: 1.087
Flammpunkt: 87 °C (188 °F)
MDL: MFCD00051495
CAS-Nummer: 30433-91-1
EINECS: 250-196-9
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR