Thiophen-2-ethylamin

Lieferant: Thermo Scientific

Synonyme: 2-(Thiophen-2-yl)ethanamine, 2-(2-Aminoethyl)thiophene

TL00533.1 TL00533.10
MAYBTL00533.1EA 0 EUR
MAYBTL00533.1 MAYBTL00533.10
Thiophen-2-ethylamin
Thiophen-2-ethylamin
Formel: C₆H₉NS
Molecular Weight: 127,21 g/mol
MDL: MFCD00051495
CAS-Nummer: 30433-91-1
EINECS: 250-196-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR