Thiophen-2-ethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Thiophen-2-yl)ethanamine, 2-(2-Aminoethyl)thiophene

M-Clarity™ Qualitätsstufe = MQ100

423270-1G 423270-5G
SIAL423270-1GEA 22.5 EUR
SIAL423270-1G SIAL423270-5G
Thiophen-2-ethylamin, Sigma-Aldrich®
Thiophen-2-ethylamin
Formel: C₆H₉NS
Molecular Weight: 127.21 g/mol
MDL: MFCD00051495
CAS-Nummer: 30433-91-1
EINECS: 250-196-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR