Thiophen-2-carbaldehydoxim ≥98%

Lieferant: Thermo Scientific
Achtung

L10961.06 L10961.14
L10961.06EA 22.6 EUR
L10961.06 L10961.14
Thiophen-2-carbaldehydoxim ≥98%
Thiophen-2-carbaldehydoxim

Purity: 98+% CAS No.: 29683-84-9 Molecular Formula: C5H5NOS Formula Weight: 127.17 Melting Point: 132-136° Beilstein Reference: 108676

Formel: C₅H₅NOS
Molecular Weight: 127,17 g/mol
Schmelzpunkt: 132…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022509
CAS-Nummer: 29683-84-9
EINECS: 249-778-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR