Thiophen-3-carbaldehydoxim

Lieferant: CHEMPUR

RF01738.250 RF01738.1
CHMPRF01738.250EA 0 EUR
CHMPRF01738.250 MAYBRF01738.1
Thiophen-3-carbaldehydoxim
Thiophen-3-carbaldehydoxim
Formel: C₅H₅NOS
Molecular Weight: 127,17 g/mol
MDL: MFCD00151853
CAS-Nummer: 42466-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR