N-Fmoc-glycin

Lieferant: BACHEM BIOCHEMICA

Synonyme: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)acetic acid, Fmoc-Gly-OH

4003078.0025 4003078.0100 4003078.0500
BACE4003078.0025EA 0 EUR
BACE4003078.0025 BACE4003078.0100 BACE4003078.0500
N-Fmoc-glycin
N-Fmoc-glycin
Formel: C₁₇H₁₅NO₄
Molecular Weight: 297,3 g/mol
MDL: MFCD00037140
CAS-Nummer: 29022-11-5
EINECS: 249-373-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR