4-Brom-2,1,3-benzothiadiazol

Lieferant: Thermo Scientific

Synonyme: 4-Bromobenzo[c][1,2,5]thiadiazole

CC09110EA CC09110DA
MAYBCC09110EAEA 0 EUR
MAYBCC09110EA MAYBCC09110DA
4-Brom-2,1,3-benzothiadiazol
4-Bromo-2,1,3-benzothiadiazole
Formel: C₆H₃BrN₂S
Molecular Weight: 215,0704 g/mol
MDL: MFCD00614355
CAS-Nummer: 22034-13-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR