5-Fluor-2,1,3-benzothiadiazol
Lieferant: FLUOROCHEM
Synonyme:
5-Fluorobenzo[c][1,2,5]thiadiazole
008321-1G
008321-5G
FLUO008321-1GEA
123
EUR
FLUO008321-1G
FLUO008321-5G
5-Fluor-2,1,3-benzothiadiazol
5-Fluoro-2,1,3-benzothiadiazole
Formel:
C₆H₃FN₂S Molecular Weight: 154,1700 g/mol |
MDL:
MFCD00173946 CAS-Nummer: 17821-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...