5-Fluor-2,1,3-benzothiadiazol

Lieferant: Thermo Scientific

Synonyme: 5-Fluorobenzo[c][1,2,5]thiadiazole

RDP00058.1 RDP00058.10
MAYBRDP00058.1EA 0 EUR
MAYBRDP00058.1 MAYBRDP00058.10
5-Fluor-2,1,3-benzothiadiazol
5-Fluoro-2,1,3-benzothiadiazole
Formel: C₆H₃FN₂S
Molecular Weight: 154,1700 g/mol
MDL: MFCD00173946
CAS-Nummer: 17821-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR