5-Chlor-6-fluor-2,1,3-benzothiadiazol

Lieferant: Thermo Scientific

RDP00065.250 RDP00065.1
MAYBRDP00065.250EA 0 EUR
MAYBRDP00065.250 MAYBRDP00065.1
5-Chlor-6-fluor-2,1,3-benzothiadiazol
5-Chloro-6-fluoro-2,1,3-benzothiadiazole
Formel: C₆H₂ClFN₂S
CAS-Nummer: 175204-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR