2,1,3-Benzothiadiazol-5-carbonylchlorid

Lieferant: FLUOROCHEM

CFF55.1 CFF55.5 017047-1G 017047-5G
CHMPCFF55.1EA 321 EUR
CHMPCFF55.1 CHMPCFF55.5 FLUO017047-1G FLUO017047-5G
2,1,3-Benzothiadiazol-5-carbonylchlorid
2,1,3-Benzothiadiazol-5-carbonylchlorid
Formel: C₇H₃ClN₂OS
MDL: MFCD02681992
CAS-Nummer: 321309-31-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR