2,1,3-Benzothiadiazol-5-carbonsäure
Lieferant: FLUOROCHEM
CFF54.2.5
CFF54.10
017046-2.5G
017046-10G
017046-1G
CHMPCFF54.2.5EA
481
EUR
CHMPCFF54.2.5
CHMPCFF54.10
FLUO017046-2.5G
FLUO017046-10G
FLUO017046-1G
2,1,3-Benzothiadiazol-5-carbonsäure
2,1,3-Benzothiadiazole-5-carboxylic acid
Formel:
C₇H₄N₂O₂S |
MDL:
MFCD01647555 CAS-Nummer: 16405-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...