4-Amino-5-fluorchinazolin

Lieferant: FLUOROCHEM

003992-1G 003992-5G
FLUO003992-1GEA 345 EUR
FLUO003992-1G FLUO003992-5G
4-Amino-5-fluorchinazolin
4-Amino-5-fluoroquinazoline
Formel: C₈H₆FN₃
MDL: MFCD00153064
CAS-Nummer: 137553-48-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR