2,4-Diamino-6-nitrochinazolin 98%

Lieferant: Thermo Scientific
Achtung

L12330.03 L12330.06
L12330.03EA 32.9 EUR
L12330.03 L12330.06
2,4-Diamino-6-nitrochinazolin 98%
2,4-Diamino-6-nitroquinazoline

Purity: 98% CAS No.: 7154-34-9 Molecular Formula: C8H7N5O2 Formula Weight: 205.18 Melting Point: 360° Beilstein Reference: 749531

Formel: C₈H₇N₅O₂
Molecular Weight: 205,18 g/mol
Schmelzpunkt: >360 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023910
CAS-Nummer: 7154-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR