3-Chlor-4-phenyl-1,2,5-thiadiazol

Lieferant: BIONET RESEARCH

5K-360S.500 5K-360S.1 5K-360S.5 5K-360S.10
BIOT5K-360S.500EA 0 EUR
BIOT5K-360S.500 BIOT5K-360S.1 BIOT5K-360S.5 BIOT5K-360S.10
3-Chlor-4-phenyl-1,2,5-thiadiazol
3-Chloro-4-phenyl-1,2,5-thiadiazole
Formel: C₈H₅ClN₂S
MDL: MFCD01935960
CAS-Nummer: 5728-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR