2,2'-Bithiophen-5-boronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-([2,2'-Bithiophen]-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
M-Clarity™ Qualitätsstufe = MQ100
578878-1G
578878-5G
SIAL578878-1GEA
157
EUR
SIAL578878-1G
SIAL578878-5G
2,2'-Bithiophen-5-boronsäurepinakolester, Sigma-Aldrich®
2,2'-Bithiophen-5-boronsäurepinakolester
Formel:
C₁₄H₁₇BO₂S₂ Molecular Weight: 292,2 g/mol |
MDL:
MFCD04039974 CAS-Nummer: 479719-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...