2,2'-Bithiophen-5-boronsäure
Lieferant: CHEMPUR
Synonyme:
[2,2'-Bithiophen]-5-ylboronic acid
CC14612.250
CC14612.1
CHMPCC14612.250EA
0
EUR
CHMPCC14612.250
CHMPCC14612.1
2,2'-Bithiophen-5-boronsäure
2,2'-Bithiophen-5-boronsäure
Formel:
C₈H₇BO₂S₂ Molecular Weight: 210,0810 g/mol |
MDL:
MFCD03086112 CAS-Nummer: 132898-95-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...