Tri-tert-butylphosphin 1 mol/l in Toluol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tris(1,1-dimethylethyl)phosphin
M-Clarity™ Qualitätsstufe = MQ100
655325-10ML
655325-50ML
SIAL655325-10MLEA
132
EUR
SIAL655325-10ML
SIAL655325-50ML
Tri-tert-butylphosphin 1 mol/l in Toluol, Sigma-Aldrich®
Tri-tert-butylphosphin
Formel:
C₁₂H₂₇P Molecular Weight: 202,32 g/mol |
MDL:
MFCD00015006 CAS-Nummer: 13716-12-6 EINECS: 237-266-4 UN: 2845 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...