6,8-Dimethyl-3-formylchromon

Lieferant: CHEMPUR

FL12010.POR 012010-5G
CHMPFL12010.POREA 0 EUR
CHMPFL12010.POR FLUO012010-5G
6,8-Dimethyl-3-formylchromon
6,8-Dimethyl-3-formylchromon
Formel: C₁₂H₁₀O₃
MDL: MFCD00192182
CAS-Nummer: 42059-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR