5-Chlor-6-methyl-2,1,3-benzothiadiazol

Lieferant: Thermo Scientific

RDP00248.1 RDP00248.10
MAYBRDP00248.1EA 0 EUR
MAYBRDP00248.1 MAYBRDP00248.10
5-Chlor-6-methyl-2,1,3-benzothiadiazol
5-Chloro-6-methyl-2,1,3-benzothiadiazole
Formel: C₇H₅ClN₂S
CAS-Nummer: 50636-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR