4-Methyl-2,1,3-benzothiadiazol

Lieferant: Thermo Scientific

Synonyme: 4-Methylbenzo[c][1,2,5]thiadiazole

RDP00094.250 RDP00094.1
MAYBRDP00094.250EA 0 EUR
MAYBRDP00094.250 MAYBRDP00094.1
4-Methyl-2,1,3-benzothiadiazol
4-Methyl-2,1,3-benzothiadiazole
Formel: C₇H₆N₂S
Molecular Weight: 150,2009 g/mol
MDL: MFCD00126471
CAS-Nummer: 1457-92-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR