3-Methylsalicylaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-3-methylbenzaldehyde

M-Clarity™ Qualitätsstufe = MQ100

444553-1G 444553-5G
SIAL444553-1GEA 32.3 EUR
SIAL444553-1G SIAL444553-5G
3-Methylsalicylaldehyd, Sigma-Aldrich®
3-Methylsalicylaldehyd
Formel: C₈H₈O₂
Molecular Weight: 136,1479 g/mol
MDL: MFCD00040798
CAS-Nummer: 824-42-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR