Rhodium(II)acetat dimer ≥99,99% (Metall-Basis) Rh ≥ 46.2%, Premion®
Lieferant: Thermo Scientific
Synonyme: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)
10560.01 10560.03 10560.06
10560.01EA 297 EUR
10560.01 10560.03 10560.06
Rhodium(II)acetat dimer ≥99,99% (Metall-Basis) Rh ≥ 46.2%, Premion®
Rhodium(II)acetat dimer
Purity: 99.99% CAS No.: 15956-28-2 Molecular Formula: C8H12O8Rh2 Formula Weight: 441.99 Physical Form: Powder Appearance: Green Odour: Odourless Melting Point: 205° Solubility: Slightly soluble in ethanol MDL No.: MFCD00003538
Cyclopropanation of alkenes
Formel: C₈H₁₂O₈Rh₂
Molecular Weight: 441,987 g/mol
Schmelzpunkt: 205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003538
CAS-Nummer: 15956-28-2
EINECS: 240-084-8