2-Isopropylphenylboronsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Isopropylbenzeneboronic acid, (2-Isopropylphenyl)boronic acid

L20110.03 L20110.06
L20110.03EA 103 EUR
L20110.03 L20110.06
2-Isopropylphenylboronsäure ≥97%
2-Isopropylphenylboronsäure

Purity: 97% CAS No.: 89787-12-2 Molecular Formula: C9H13BO2 Formula Weight: 164.01 Melting Point: 85-86° Beilstein Reference: 3090298

Formel: C₉H₁₃BO₂
Molecular Weight: 164,01 g/mol
Schmelzpunkt: 85…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411937
CAS-Nummer: 89787-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR