2-Isopropylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Isopropylbenzeneboronic acid, (2-Isopropylphenyl)boronic acid

BB-2201-5G BB-2201-1G
COBBBB-2201-5GEA 210 EUR
COBBBB-2201-5G COBBBB-2201-1G
2-Isopropylphenylboronsäure
2-Isopropylphenylboronsäure
Formel: C₉H₁₃BO₂
Molecular Weight: 164,01 g/mol
MDL: MFCD03411937
CAS-Nummer: 89787-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR