3-Isopropylphenylboronsäure ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Isopropylbenzeneboronic acid

L15530.03 L15530.MD
L15530.03EA 119 EUR
L15530.03 L15530.MD
3-Isopropylphenylboronsäure ≥99%
3-Isopropylphenylboronsäure

Purity: 99% CAS No.: 216019-28-2 Molecular Formula: C9H13BO2 Formula Weight: 164.01 Flash Point: 110°(230°F) Melting Point: ca 75-80° Beilstein Reference: 3090001 MDL No.: MFCD01074634

Formel: C₉H₁₃BO₂
Molecular Weight: 164,01 g/mol
Schmelzpunkt: ∼ 75…80 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD01074634
CAS-Nummer: 216019-28-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR