3-Isopropylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Isopropylbenzeneboronic acid
BB-2133-5G
BB-2133-25G
BB-2133-1G
COBBBB-2133-5GEA
124
EUR
COBBBB-2133-5G
COBBBB-2133-25G
COBBBB-2133-1G
3-Isopropylphenylboronsäure
3-Isopropylphenylboronsäure
Formel:
C₉H₁₃BO₂ Molecular Weight: 164,01 g/mol |
MDL:
MFCD01074634 CAS-Nummer: 216019-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...