4-Isopropylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Isopropylbenzeneboronic acid
BB-2132-5G
BB-2132-25G
BB-2132-1G
COBBBB-2132-5GEA
66
EUR
COBBBB-2132-5G
COBBBB-2132-25G
COBBBB-2132-1G
4-Isopropylphenylboronsäure
4-Isopropylphenylboronsäure
Formel:
C₉H₁₃BO₂ Molecular Weight: 164,01 g/mol |
MDL:
MFCD01074614 CAS-Nummer: 16152-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...