4-Brom-3-(trifluormethyl)anisol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-5-methoxybenzotrifluoride, 4-Bromo-3-trifluoromethylanisole
OT-0668-25G
OT-0668-1G
OT-0668-5G
COBBOT-0668-25GEA
175
EUR
COBBOT-0668-25G
COBBOT-0668-1G
COBBOT-0668-5G
4-Brom-3-(trifluormethyl)anisol
4-Brom-3-(trifluormethyl)anisol
Formel:
C₈H₆BrF₃O Molecular Weight: 255,04 g/mol |
MDL:
MFCD08059527 CAS-Nummer: 400-72-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...