3-Brom-p-anisidin
Lieferant: COMBI-BLOCKS
Synonyme:
3-Bromo-4-methoxyaniline
AN-1573-5G
AN-1573-1G
COBBAN-1573-5GEA
50.8
EUR
COBBAN-1573-5G
COBBAN-1573-1G
3-Brom-p-anisidin
3-Brom-p-anisidin
Formel:
C₇H₈BrNO Molecular Weight: 202.0485 g/mol |
MDL:
MFCD04094339 CAS-Nummer: 19056-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...