Chlorbis(2-methylphenyl)phosphin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Di-o-tolylchlorophosphine, Chlorodi(o-tolyl)phosphine

M-Clarity™ Qualitätsstufe = MQ100

686697-1G 686697-5G
SIAL686697-1GEA 91.5 EUR
SIAL686697-1G SIAL686697-5G
Chlorbis(2-methylphenyl)phosphin, Sigma-Aldrich®
Chlorbis(2-methylphenyl)phosphin
Formel: C₁₄H₁₄ClP
Molecular Weight: 248,688 g/mol
MDL: MFCD04038733
CAS-Nummer: 36042-94-1
UN: 1760
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR