Chlorbis(2-methylphenyl)phosphin ≥98%

Lieferant: Strem Chemicals, Inc.
Gefahr

Synonyme: Di-o-tolylchlorophosphine, Chlorodi(o-tolyl)phosphine

15-1815-1G 15-1815-5G
STRE15-1815-1GEA 169 EUR
STRE15-1815-1G STRE15-1815-5G
Chlorbis(2-methylphenyl)phosphin ≥98%
Chlorbis(2-methylphenyl)phosphin

Phosphine

Formel: C₁₄H₁₄ClP
Molecular Weight: 248,688 g/mol
MDL: MFCD04038733
CAS-Nummer: 36042-94-1
UN: 1760
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR