3,6-Dichlorpicolinsäure 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3,6-Dichloropyridine-2-carboxylic acid

H27146.06 H27146.03
H27146.06EA 168 EUR
H27146.06 H27146.03
3,6-Dichlorpicolinsäure 96%
3,6-Dichlorpicolinsäure

Purity: 96% CAS No.: 1702-17-6 Molecular Formula: C6H3Cl2NO2 Formula Weight: 192.00 Flash Point: 100°(212°F) Melting Point: ca 150° dec. Beilstein Reference: 473755 Merck Reference: 14,2400 MDL No.: MFCD00078655

Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192.00 g/mol
Schmelzpunkt: ∼ 150 °C (Zers.)
Flammpunkt: >100 °C (212 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00078655
CAS-Nummer: 1702-17-6
EINECS: 216-935-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR