3'-Ethinylacetanilid
Lieferant: BIONET RESEARCH
Synonyme:
N-(3-Ethynylphenyl)acetamide
7Y-0831.1
7Y-0831.10
7Y-0831.5
7Y-0831.500
BIOT7Y-0831.1EA
0
EUR
BIOT7Y-0831.1
BIOT7Y-0831.10
BIOT7Y-0831.5
BIOT7Y-0831.500
3'-Ethinylacetanilid
3'-Ethinylacetanilid
Formel:
C₁₀H₉NO Molecular Weight: 159,1846 g/mol |
MDL:
MFCD00831583 CAS-Nummer: 70933-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...